missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Grinding Vial Set, Large Vial, PC Cylinder, SS End Plugs and Impactor

Product Code. 12528696 Shop All SPEX Certiprep Products
Change view
Click to view available options
:
Large Grinding Vial Set; Capacity: 1 to 50 mL; Material: Polycarbonate, Stainless Steel
Product Code.
12528696 Large Grinding Vial Set; Capacity: 1 to 50 mL; Material: Polycarbonate, Stainless Steel
1 options
This item is not returnable. View return policy

Product Code. 12528696

Brand: Cole-Parmer™ 6801

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Large Grinding Vial Set for use with SPEX SamplePrep™ Large-Capacity Freezer/Mill Cryogenic Grinder.

  • Makes one complete grinding vial plus three spare polycarbonate center cylinders.
  • Grinds one large sample at a time.
  • Sample capacity (grinding): 1 to 50 mL
  • Typical Sample Size: 25 g
  • Withstands liquid nitrogen temperatures.
  • Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples.
  • Transparent plastic center cylinder allows visual check of grinding process.
  • Stainless steel end plugs and impactor are autoclavable and reusable.
  • Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Compatible with 6804 Large Extractor/Vial Opener.
  • Components available separately.
  • For grinding samples such as plant and animal tissue, seeds, polymers, pharmaceuticals, textiles, hair, and bones.

Specifications

Product Type Grinding Set
Capacity (English) 0.03 to 5.07 oz.
Capacity (Metric) 1 to 50 mL
For Use With (Application) Grinding Samples Such as Plant and Animal Tissue, Seeds, Polymers, Pharmaceuticals, Textiles, Hair and Bones
For Use With (Equipment) Large Extractor/Vial Opener 6804, Auto-Extractor 6814 of Spex SamplePrep Large Capacity Freezer/Mill
Includes 1 Large Stainless Steel Impactor, 2 Large Stainless Steel End Plugs, and 1 Pk. of Large Polycarbonate Center Cylinders
Material Stainless Steel
Material (Secondary) Polycarbonate
Sample Volume (English) 0.03 to 1.69 oz.
Sample Volume (Metric) 1 to 50 mL
Size Large
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.