missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sphingosine-1-phosphate
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 5 mg |
| Description | White to off-white |
| Content And Storage | -30°C to -10°C |
| Formula Weight | 379.47 |
| Sensitivity | Light Sensitive |
| Product Type | Sphingosine-1-phosphate |
| Form | Powder |
| InChI Key | DUYSYHSSBDVJSM-KRWOKUGFSA-N |
| SMILES | CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O |
| IUPAC Name | {[(2S,3R,4E)-2-amino-3-hydroxyoctadec-4-en-1-yl]oxy}phosphonic acid |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion