missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sphingosine-1-phosphate

Product Code. 30564791
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30564791 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30564791 Supplier Tocris Bioscience™ Supplier No. 1370/1
  • ONLINE EXCLUSIVE PRICE
    285.00 €
    268.80 € / 1mg
    Save 16.20 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Endogenous agonist at S1P1-5

Endogenous second messenger involved in the control of cell proliferation and motility, and Ca2+ mobilization. Acts as an agonist at sphingosine-1-phosphate receptors (S1P1-5) and as an activator of GPR3, GPR6 and GPR12. Effectors regulated include p38 MAP kinase, PLC, adenylyl cyclase, myosin light chain phosphatase and focal adhesion kinase.
TRUSTED_SUSTAINABILITY

Specifications

CAS 26993-30-6
Quantity 1 mg
Molecular Formula C18H38NO5P
Formula Weight 379.48
Purity >98%
Molecular Weight (g/mol) 379.478
Chemical Name or Material Sphingosine-1-phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.