Learn More
Heidolph™ Spider with Flasks
Product Code. 10688382 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10688382 50 mL Each
1 options
Product Code. 10688382 Supplier Heidolph™ Supplier No. 1530000504
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with rotary evaporators
Comes with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Type Glassware Assembly Rotary Evaporator
Volume (Metric) 50 mL
Voltage 115/230 VAC