Description
- Convenient adapters, enable spinning of all gravity columns.
Specifications
Specifications
| Description | Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns. |
| Material | Polypropylene |
| Quantity | 10 Pack |
| For Use With (Application) | Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD-10 desalting column for centrifugation |
| Type | Spin Column Adapter |
| Column Type | For PD-10 Desalting Column |