missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy DNA/RNA/Protein All In One Kit

Product Code. 17702448 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
50 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17702448 50 Preps 50 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17702448 Supplier MP Biomedicals™ Supplier No. 116544050
  • ONLINE EXCLUSIVE PRICE
    687.00 €
    651.00 € / 50 sets
    Save 36.00 €
    5% Off
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

SPINeasy DNA/RNA/Protein All In One Kit - Quick and efficient isolation of DNA, RNA and protein from the same sample using spin column technology.

  • SPINeasy DNA/RNA/Protein All-In-One Kit utilizes a convenient workflow and silica-membrane spin-columns to isolate without the use of toxic substances such as phenol and chloroform:
    • DNA
    • RNA
    • Protein components from the same sample.
  • The use of our specially formulated Lysis Buffer R and Lysing Matrix A in combination with FastPrep™ Instruments from MP Biomedicals enables highly efficient lysis of tissue samples within seconds. DNA, RNA and proteins are then sequentially purified from the same lysate. Briefly, DNA is adsorbed onto the first spin column, while the flow-through is collected and used for RNA purification. The second spin column captures RNA and the flow-through from this step is used for protein extraction. DNA and RNA are bound on the first and second columns, respectively, and then washed and eluted. In the final extraction, proteins are:
    • Precipitated out of solution
    • Pelleted down by centrifugation
    • Washed and resuspended.
  • Each molecular component is then immediately available for their respective downstream applications.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Kit
    Scale Mini
    Sample Type Human, Mouse, Rat
    Sequencing Type DNA and RNA Sequencing
    Quantity 50 Preps
    compliance-icons compliance-icons compliance-icons
    Product Identifier
    • SPINeasy DNA/RNA/Protein All In One Kit, 50 preps
    Signal Word
    • Danger
    Hazard Category
    • Acute toxicity Category 4
    • Serious eye damage/eye irritation Category 1
    • Serious eye damage/eye irritation Category 2
    • Skin corrosion/irritation Category 2
    • Specific target organ toxicity after repeated exposure Category 2
    Hazard Statement
    • H302-Harmful if swallowed.
    • H315-Causes skin irritation.
    • H318-Causes serious eye damage.
    • H319-Causes serious eye irritation.
    • H373-May cause damage to organs.
    Precautionary Statement
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P264-Wash thoroughly after handling.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P321-Specific treatment (see on this label).
    • P405-Store locked up.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contain: Guanidine Hydrochloride, Manganese Sulfate Monohydrate, Calcium Chloride, Zinc Sulfate heptahydrate
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.