missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals SPINeasy RNA kit for Soil

Product Code. 30281096 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Reactions:
50 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
30281096 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281096 Supplier MP Biomedicals Supplier No. 116585050
  • ONLINE EXCLUSIVE PRICE
    545.00 €
    516.60 € / Pack of 1
    Save 28.40 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SPINeasy RNA kit for Soil which utilizes silica-membrane spin-column technology for efficient and high-purity RNA extraction.

The SPINeasy RNA Kit for Soil is a high-performance RNA extraction kit utilizing silica-membrane spin-column technology.

This kit allows for the efficient isolation of RNA from various soil types, including:

  • those with high humic acid content
  • heavy contaminants
  • compost
  • gardening soil
  • and low-biomass soils.

The extraction process can be completed in under an hour with:

  • minimal RNA degradation
  • and the use of hazardous chemicals like phenol or chloroform is unnecessary.

Our specially formulated inhibitor removal technology effectively handles soil samples containing:

  • humic acids
  • heavy metals
  • and other aromatic compounds

ensuring accurate PCR results.

RNA extracted with this kit demonstrates high integrity and purity, suitable for downstream applications such as:

  • reverse transcription
  • real-time PCR
  • and sequencing.

Usage Statement: Research Use Only (RUO)

Specifications

Sample Type Soil
No. of Reactions 50 Preps
Product Type RNA Isolation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.