missing translation for 'onlineSavingsMsg'
Learn More

Spinetoram, TRC

Product Code. 17941455
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941455 10 mg 10mg
17983835 1 mg 1mg
17961775 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17941455 Supplier Toronto Research Chemicals Supplier No. TRCS68365310MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187166-40-1
Molecular Formula C42 H69 N O10
Molecular Weight (g/mol) 748
Synonym Spinetoram, 1H-as-Indaceno[3,2-d]oxacyclododecin-7,15-dione, 2-[(6-deoxy-3-O-ethyl-2,4-di-O-methyl-α-L-mannopyranosyl)oxy]-13-[[(2R,5S,6R)-5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]oxy]-9-ethyl-2,3,3a,4,5,5a,5b,6,9,10,11,12,13,14,16a,16b-hexadecahydro-14-methyl-, (2R,3aR,5aR,5bS,9S,13S,14R,16aS,16bR)-, 1H-as-Indaceno[3,2-d]oxacyclododecin-7,15-dione, 2-[(6-deoxy-3-O-ethyl-2,4-di-O-methyl-α-L-mannopyranosyl)oxy]-13-[[(2R,5S,6R)-5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]oxy]-9-ethyl-2,3,3a,4,5,5a,5b,6,9,10,11,12,13,14,16a,16b-hexadecahydro-14-methyl-, [2R-(2R*,3aR*,5aR*,5bS*,9S*,13S*,14R*,16aS*,16bR*)]-, (2R,3aR,5aR,5bS,9S,13S,14R,16aS,16bR)-2-[(6-Deoxy-3-O-ethyl-2,4-di-O-methyl-α-L-mannopyranosyl)oxy]-13-[[(2R,5S,6R)-5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]oxy]-9-ethyl-2,3,3a,4,5,5a,5b,6,9,10,11,12,13,14,16a,16b-hexadecahydro-14-methyl-1H-as-indaceno[3,2-d]oxacyclododecin-7,15-dione, DE 175J, Spinetoram J, Spinetoram, DE 175J, Spinetoram J
SMILES CCO[C@@H]1[C@@H](OC)[C@H](C)O[C@@H](O[C@@H]2C[C@H]3CC[C@H]4[C@@H]5CC(=O)O[C@@H](CC)CCC[C@H](O[C@H]6CC[C@@H]([C@@H](C)O6)N(C)C)[C@@H](C)C(=O)C5=C[C@H]4[C@@H]3C2)[C@@H]1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C42H69NO10/c1-10-27-13-12-14-35(53-37-18-17-34(43(6)7)24(4)49-37)23(3)38(45)33-21-31-29(32(33)22-36(44)51-27)16-15-26-19-28(20-30(26)31)52-42-41(47-9)40(48-11-2)39(46-8)25(5)50-42/h21,23-32,34-35,37,39-42H,10-20,22H2,1-9H3/t23-,24-,25+,26-,27+,28-,29-,30-,31-,32+,34+,35+,37+,39+,40-,41-,42+/m1/s1
Formula Weight 747.4921
Percent Purity >95%
Chemical Name or Material Spinetoram J
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.