missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Spinosad, mixture of A and D

Product Code. 17261316 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17261316 5 g 5g
17251316 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17261316 Supplier Thermo Scientific Chemicals Supplier No. 467190050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 168316-95-8 | C42H71NO9 | 734.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168316-95-8

Specifications

Quantity 5 g
Melting Point 142.0°C to 145.0°C
Molecular Formula C42H71NO9
InChI Key RQOIAWYOVOXMST-UHFFFAOYNA-N
SMILES CCC1CCCC(OC2CCC(C(C)O2)N(C)C)C(C)C(=O)C2CC3C4CC(CC4CCC3C2CC(=O)O1)OC1CC(C)C(OC)C(OC)C1OC
IUPAC Name 13-{[5-(dimethylamino)-6-methyloxan-2-yl]oxy}-9-ethyl-14-methyl-2-[(2,3,4-trimethoxy-5-methylcyclohexyl)oxy]-icosahydro-1H-as-indaceno[3,2-d]oxacyclododecane-7,15-dione
Molecular Weight (g/mol) 734.03
Chemical Name or Material Spinosad

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.