missing translation for 'onlineSavingsMsg'
Learn More

Spinosad, TRC

Product Code. 30012794
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012794 25 mg 25mg
30003637 5 mg 5mg
30004718 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012794 Supplier Toronto Research Chemicals Supplier No. TRCS68192025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168316-95-8
Molecular Formula C42 H67 N O10 . C41 H65 N O10
Molecular Weight (g/mol) 747
Synonym Blackhawk, Comfortis, Conserve, Entrust, Extinosad, GF 120, GF 120 Fruit Fly Bait, GF 120 NF Naturalyte, GF 120 Naturalyte Fruit Fly Bait, GF 1587, Gardens Alive Bulls-Eye, LY 232105, Laser, Laser (insecticide), Naturalyte, PP 105, SpinTor, Spinoace, Spinosad NT, Spinosyn A-spinosyn D mixt., Success, Success 4, Success Naturalyte, Tracer, Tracer Naturalyte, Tracer Naturalyte Insect Control, XDE 105
IUPAC Name (1S,2S,5R,7S,9S,10S,14R,15S,19S)-15-[(2R,5S,6R)-5-(dimethylamino)-6-methyloxan-2-yl]oxy-19-ethyl-4,14-dimethyl-7-[(2R,3R,4R,5S,6S)-3,4,5-trimethoxy-6-methyloxan-2-yl]oxy-20-oxatetracyclo[10.10.0.02,10.05,9]docosa-3,11-diene-13,21-dione
SMILES CC[C@H]1CCC[C@H](O[C@H]2CC[C@@H]([C@@H](C)O2)N(C)C)[C@@H](C)C(=O)C3=C[C@H]4[C@@H]5C[C@@H](C[C@H]5C(=C[C@H]4[C@@H]3CC(=O)O1)C)O[C@@H]6O[C@@H](C)[C@H](OC)[C@@H](OC)[C@H]6OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C42H67NO10/c1-11-26-13-12-14-35(53-37-16-15-34(43(6)7)24(4)49-37)23(3)38(45)33-20-31-29(32(33)21-36(44)51-26)17-22(2)28-18-27(19-30(28)31)52-42-41(48-10)40(47-9)39(46-8)25(5)50-42/h17,20,23-32,34-35,37,39-42H,11-16,18-19,21H2,1-10H3/t23-,24-,25+,26+,27-,28+,29-,30-,31-,32+,34+,35+,37+,39+,40-,41-,42+/m1/s1
Formula Weight 745.4765
Percent Purity >95%
Chemical Name or Material Spinosad
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.