Learn More
Lenz Laborglasintrumente Spiral Condensers, with GL Threads
Description
- Spiral condensers for cooling condensates
- With NS socket and NS cone
- Water connection: GL 14 threads (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
Specifications
| Includes | GL 14 threads and two threaded plastic olives |
| Length (Metric) | 400 mm |
| Standard Taper Joint No | Socket and Cone NS 29/32 |