missing translation for 'onlineSavingsMsg'
Learn More

Spiro-(1,3-dioxolane-2,3'indolin)-2'-one, 97%, Thermo Scientific™

Product Code. 10624775
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10624775 25 g 25g
10180692 1 g 1g
10190692 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10624775 Supplier Thermo Scientific Maybridge Supplier No. S13949EB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6714-68-7
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00173689
InChI Key PRMHWSVVQZVDGR-UHFFFAOYSA-N
Synonym spiro 1,3-dioxolane-2,3'-indol-2' 1'h-one, spiro-1,3-dioxolane-2,3'indolin-2'-one, spiro 1,3 dioxolane-2,3'-indolin-2'-one, spiro 1,3-dioxolane-2,3'-3h indol-2' 1'h-one, 1'h-spiro 1,3-dioxolane-2,3'-indole-2'-one, spiro 1,3-dioxolane-2,3'-indoline-7-one, 1'h-spiro 1,3-dioxolane-2,3'-indol-2'-one, maybridge1_007498, chembl6816
PubChem CID 281156
IUPAC Name spiro[1,3-dioxolane-2,3'-1H-indole]-2'-one
SMILES C1COC2(O1)C3=CC=CC=C3NC2=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material Spiro-(1, 3-dioxolane-2, 3'indolin)-2'-one
compliance-icons
Product Identifier
  • Spiro-(1,3-dioxolane-2,3'indolin)-2'-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.