missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone, MedChemExpress

Product Code. 16480998
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480998 5 g 5g
16412239 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480998 Supplier Medchem Express Supplier No. HYB0561
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Spironolactone (SC9420) is an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. Spironolactone is also a potent antagonist of androgen receptor with an IC50 of 77 nM. Spironolactone promotes autophagy in podocytes.

Spironolactone (SC9420) is an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. Spironolactone is also a potent antagonist of androgen receptor with an IC50 of 77 nM. Spironolactone promotes autophagy in podocytes.

Specifications

Chemical Name or Material Spironolactone
CAS 52-01-7
Color White
Purity Grade Notes Research
Molecular Formula C24H32O4S
Quantity 5 g
Synonym SC9420
Solubility Information DMSO : ≥ 50 mg/mL (120.03 mM) ∣H2O : 0.1 mg/mL (0.24 mM; Need ultrasonic)
SMILES C[C@@]12[C@](OC3=O)(CC3)CC[C@@]1([H])[C@@]([C@@H](CC4=CC5=O)SC(C)=O)([H])[C@]([C@]4(CC5)C)([H])CC2
Molecular Weight (g/mol) 416.57
Formula Weight 416.57
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.