missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J60119.06

Quantity 5g

 View more versions of this product

Product Code. 15412949

  • 121.00 € / 5g
 Restricted Item
Estimated Shipment: 15-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

52-01-7
416.576
LXMSZDCAJNLERA-ZHYRCANASA-N
CHEBI:9241
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
C24H32O4S
MFCD00082250
5833
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate

Specifications

52-01-7
5g
MFCD00082250
Soluble in chloroform and methanol. Slightly soluble in water.
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
416.576
CHEBI:9241
Spironolactone
White
C24H32O4S
14,8760
LXMSZDCAJNLERA-ZHYRCANASA-N
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
5833
416.58
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.