Learn More
Cytiva Splash Guard for Delivery Port
Product Code. 10536545 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Splash Guard
Unit Size:
Each
Product Code. Product Type Unit Size
10536545 Splash Guard Each
1 options
Product Code. 10536545 Supplier Cytiva Supplier No. 80634201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
For Use With (Application) For delivery port, prevents splashing of reagents into staining tray
Product Type Splash Guard