missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Spoked Ratchet Nut for FastPrep-24™ Classic

Product Code. 11411535 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-24™ Classic
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11411535 FastPrep-24™ Classic Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411535 Supplier MP Biomedicals™ Supplier No. 116004510
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spoked Ratchet Nut for FastPrep-24™ Classic is a replacement tightening button used to fix adapters on both FastPrep-24 Classic.

  • Securing knob for FastPrep-24™ used to lock the tube holder on 3 step assembly

Specifications

For Use With (Application) Replacement securing knob used to lock the adapter on 3 step assembly.
For Use With (Equipment) FastPrep-24™ Classic
Packaging Quantity 2 Pk.

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.