missing translation for 'onlineSavingsMsg'
Learn More

Steris™ Spor Klenz RTU Euro

Product Code. 15396810
Change view
Click to view available options
Volume (Metric):
850 mL
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15396810 850 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15396810 Supplier Steris™ Supplier No. 6525M2EC
  • ONLINE EXCLUSIVE PRICE
    368.00 €
    246.67 € / Pack of 4
    Save 121.33 €
    32% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Convenient, ready-to-use cold sterilant specifically formulated for use on hard environmental surfaces. Steris™ Spor Klenz RTU Euro is a stabilized blend of chemicals that provides fast, effective microbial control.

  • Fast, effective microbial control against bacteria, fungi, viruses, and bacteria spores proven using AOAC and EN protocols.
  • Suitable for use as a sterilant, sporicide, disinfectant or sanitizer.
  • Filtered to 0.22 micron.
  • Suitable for steriizing and disinfecting hard surfaces.
  • Suitable for use in pharmaceutical, medical device, biotech, and cosmetic manufacturing facilities.
  • All active ingredients are notified according to the European Union Biocidal Products Directive (BPD).
  • Low toxicity profile.
  • Requires no mixing or activation and is reusable for up to 14 days.
  • Each lot is fully traceable and documented by a Certificate of Analysis including filtration and USP sterility.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sporicide/Disinfectant
For Use With (Application) Microbial control against bacteria, fungi, viruses, and bacteria spores proven using AOAC and EN protocols.
Quantity 4/Cs.
Volume (Metric) 850 mL
compliance-icons
Product Identifier
  • Spor-Klenz Ready to Use Sporicide/Disinfectant
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid, Hydrogen peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.