Learn More
uvex 9193 Sportstyle Safety Spectacles
Product Code. 17787689 Shop All Uvex Products
Change view
Click to view available options
Supravision ETC, Clear
Product Code.
17787689 Supravision ETC, Clear
1 options
Product Code. 17787689 Supplier uvex Supplier No. 9193415
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 06-08-2026
Add to basket
Edge
Add to basket
Innovative safety spectacles featuring a sporty design, combined with maximum functionality and safety. Feather-light construction weighs only 23 grams, making these spectacles comfortable to wear for extended periods of time.
Modern eye protection. The uvex sportstyle stands for a new generation of innovative safety glasses: a clean, sporty design combining maximum functionality and protection – these lightweight safety glasses are a joy for everyone.
General features
  • Sporty, lightweight safety glasses with contoured lens for a great fit on narrow or wider faces
  • Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
  • Frame color: anthracite, blue
  • PC lens: clear
Protection features
  • Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
  • Marking: W 166 FT CE – 2C-1.2 W1 FTKN CE
  • UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
Comfort features
  • The combination of soft padding around the forehead and ergonomically shaped arms with soft ear pieces ensures a comfortable and secure fit without creating pressure points
  • Soft, adjustable nose piece makes it possible to adjust the glasses to the wearer and ensures a comfortable, non-slip, pressure-free fit
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Supravision ETC
Lens Coating Anti-fog
Color Anthracite, Blue
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard