missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex Sportstyle Planet Clear Spectacles
Shop All Uvex Products

Click to view available options
:
Lens Tint: Clear
Description
- Modern eye protection
- Feature an innovative close-fitting, sporty design that suits everyone
- The soft padding around the forehead and on the ear-pieces provides a comfortable and secure grip
- Their lightweight, ergonomic design offers maximum wearer comfort even when worn for long periods
- You hardly notice that you are wearing safety spectacles, but you still have optimum protection

Specifications
Specifications
Lens Coating | Uncoated |
Lens Tint | Clear |
Product Type | Safety Glasses |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Polypropylene, TPE |
UV Protection | UV400 |
Frame Type | Sport |
Autoclavable | Not Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction