Learn More
Tuttnauer Spray Arm Monitor Chamber Sensor
Product Code. 17868309 Shop All Tuttnauer Products
Click to view available options
:
Spray Arm Monitor; For use with: Tiva Washer
Product Code. 17868309
Brand: Tuttnauer 119904311
Only null left
Add to basket
Add to basket
The Spray Arm Monitor Chamber Sensor is a special sensor to control the frequency of spray arm rotation inside the washing chamber. If the spray arm is blocked, the operator is alerted by an alarm. This ensures that all glassware is washed thoroughly.
  • Special sensor to control the frequency of spray arm rotation inside the washing chamber.
  • If the spray arm is blocked, the operator is alerted by an alarm.
  • This ensures that all glassware is washed thoroughly.
Specifications
Material Stainless Steel
Product Type Sensor
For Use With (Equipment) Tiva Washer
Quantity 1 Ea.