missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Spring Clamps for Mighty Small™ Electrophoresis Units

Product Code. 10608105
Change view
Click to view available options
For Use With (Equipment):
SE 250, SE 260 Mini Vertical Electrophoresis Unit
SE 250, SE 260 Mini Vertical Electrophoresis Unit, SE 615, SE 675 Multiple Gel Casters
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10608105 SE 250, SE 260 Mini Vertical Electrophoresis Unit Pack of 4
11560724 SE 250, SE 260 Mini Vertical Electrophoresis Unit, SE 615, SE 675 Multiple Gel Casters Pack of 4
12805856 SE 250, SE 260 Mini Vertical Electrophoresis Unit, SE 615, SE 675 Multiple Gel Casters Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10608105 Supplier Hoefer™ Supplier No. SE252
  • ONLINE EXCLUSIVE PRICE
    82.15 €
    77.24 € / Pack of 4
    Save 4.91 €
    5% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Mighty Small electrophoresis units

  • Multiple Gel Casters Replacement Parts
  • Some are Asymetrical Clamps

Specifications

Color Red
For Use With (Equipment) SE 250, SE 260 Mini Vertical Electrophoresis Unit
Type Spring Clamp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.