Learn More
Edulab Spring Extension, Steel, Pack of 200
Product Code. 30280265
Change view
Click to view available options
Product Type:
Replacement Spring
Unit Size:
Pack of 200
Product Code. Product Type Unit Size
30280265 Replacement Spring Pack of 200
1 options
Product Code. 30280265 Supplier Edulab Supplier No. VTN12302388
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Coiled steel spring with double loop ends, ideal for physics experiments and motion studies.
  • Steel coiled spring with double loop ends for secure attachment
  • End-to-end length of 50 mm (1.96 in.) and collapsed length of 24 mm (0.94 in.) without hoops
  • Outside diameter of 15 mm (0.59 in.)
  • Extends approximately 200-230 mm (8-9 in.) and returns to original length without deformity
  • Ideal for studying motion of mass on a spring, Hooke’s Law, potential energy, and related physics concepts
  • Provides a visually and kinesthetically effective method for learning spring dynamics
  • Single spring included for individual experiments
Specifications
Product Type Replacement Spring