missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRTN Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00 € - 468.00 €
Specifications
| Antigen | SPRTN |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SPRTN Polyclonal antibody specifically detects SPRTN in Human samples. It is validated for Western BlotSpecifications
| SPRTN | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C1orf124, chromosome 1 open reading frame 124, DDDL1880, dJ876B10.3, DKFZP547N043, PRO4323, RP5-876B10.3, Spartan, SprT-like N-terminal domain, zinc finger RAD18 domain-containing protein C1orf124 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human SPRTN (NP_001010984.1). MDDDLMLALRLQEEWNLQEAERDHAQESLSLVDASWELVDPTPDLQALFVQFNDQFFWGQLEAVEVKWSVRMTLCAGICSYEGKGGMCSIRLSEPLLKLRPRKDLVETLLHEMIHAYLFVTNNDKDREGHGPEFCKHMHRINSLTGANITVYHTFHDEVDEYRRHWWRCNGPCQHRPPYYGYVKRATNREPSAHDYWWAEHQKTCGGTYIKIKEPENYSKKGKGKAKLGKEPVLAAENKG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 83932 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title