missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SPT Imidazopyridine 1
GREENER_CHOICE

Product Code. p-7116095
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18760809 10 mg 10mg
18792469 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18760809 Supplier Tocris Bioscience™ Supplier No. 6055/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent serine palmitoyl transferase inhibitor

SPT Imidazopyridine 1 is a potent serine palmitoyl transferase (SPT) inhibitor (IC50 = 5 nM). Dose-dependently inhibits incorporation of 14C-serine into ceramide. Reduces plasma ceramide levels in vivo. Also increases plasma HDL and reduces vLDL cholesterol levels in rats.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[6-(1-Oxoheptyl)imidazo[1,2-a]pyridin-8-yl]benzeneacetic acid
CAS 1933533-18-6
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C22H24N2O3
Formula Weight Observed MW: 364.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.