missing translation for 'onlineSavingsMsg'
Learn More

Squalane, ≥99.0% (GC), Solstice

Product Code. 15647350
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15647350 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15647350 Supplier Honeywell-Fluka Supplier No. 8562950ML
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

for GC,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-01-3
Melting Point -38°C
Color Colorless
Density 0.81g/mL at 25°C
pH Neutral
Boiling Point 176°C (0.07 hPa)
Flash Point 218°C (424.4°F)
Odor Odorless
Packaging Glass bottle
Molecular Formula C30H62
Linear Formula [(CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)CH2CH2-]2
MDL Number MFCD00008953
Quantity 50 mL
UN Number NONH for all modes of transport
Beilstein 776019
Synonym squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
Solubility Information Insoluble in water
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYSA-N
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
Molecular Weight (g/mol) 422.826
PubChem CID 8089
Formula Weight 422.81g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material Squalane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.