missing translation for 'onlineSavingsMsg'
Learn More

Squalane, 99%, Thermo Scientific Chemicals

Product Code. 10194790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194790 100 mL Glass bottle 100mL
10002990 500 mL Glass bottle 500mL
11335922 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10194790 Supplier Thermo Scientific Acros Supplier No. 215351000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-01-3
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYSA-N
Synonym squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
PubChem CID 8089
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

Specifications

Acidity 0.2mg KOH/g max.
Melting Point 0.3°C
Color Colorless
Density 0.8085g/mL
Boiling Point 210°C to 215°C (1.0 mmHg)
Flash Point 217°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C30H62
Refractive Index 1.4510 to 1.4525
Quantity 100 mL
Saponification Value 1mg KOH/g max.
Beilstein 01,I,72
Merck Index 15,8895
Specific Gravity 0.8085
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in glacial acetic acid,readily soluble in ether,gasoline,benzene,,petroleum ether,chloroform and oils,slightly soluble in methanol,ethanol and acetone
Molecular Weight (g/mol) 422.82
Viscosity 48.2 mPa.s (20°C)
Formula Weight 422.82
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Squalane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.