missing translation for 'onlineSavingsMsg'
Learn More

Squalane, 99%, Thermo Scientific Chemicals

Product Code. 11335922 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11335922 2.5 L Glass bottle 2.5 L
10194790 100 mL Glass bottle 100mL
10002990 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335922 Supplier Thermo Scientific Acros Supplier No. 215350025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-01-3
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYSA-N
Synonym squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
PubChem CID 8089
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

Specifications

Acidity 0.2mg KOH/g max.
Color Colorless
Density 0.8085g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C30H62
Refractive Index 1.4510 to 1.4525 (20°C, 589nm)
Quantity 2.5 L
Saponification Value 1mg KOH/g max.
Molecular Weight (g/mol) 422.826
Formula Weight 422.82
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material Squalane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.