missing translation for 'onlineSavingsMsg'
Learn More

Squalane, TRC

Product Code. 30417134
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417134 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417134 Supplier Toronto Research Chemicals Supplier No. TRCS68375050G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Squalane
CAS 111-01-3
Purity Grade Notes GC
Molecular Formula C30 H62
Quantity 50 g
Synonym Tetracosane','2,6,10,15,19,23-hexamethyl- (6CI','8CI','9CI','ACI), 2,6,10,15,19,23-Hexamethyltetracosane (ACI), Biolip P 90, Cetiol SQ, Cosbiol, Dodecahydrosqualene, Fitoderm, Mild Finish 20P, Neossance Squalane, Neossance squalene, Nikko Squalane, Nikkol Refined Olive Squalane, Nikkol Sugar Squalane, NSC 6851, Perhydrosqualene, Phtyosqualan, Phytiane LS, Phytolane LS, Phytosqualan, Phytosqualane, Plantasens Olive Squalane, Pripure 3759, Pripure 379, Pripure SQV 3759, Robane, Sophim Squalane-S, Spinacane, SQ-CONO, Squalan, Squalane, Squalane N, Sugar Squalane, Super Squalane, Vitabiosol
InChI Formula InChI=1S/C30H62/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h25-30H,9-24H2,1-8H3
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
Molecular Weight (g/mol) 422.81
Formula Weight 422.4852
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.