missing translation for 'onlineSavingsMsg'
Learn More

Squalene, 98%, Thermo Scientific Chemicals

Product Code. 11410007
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410007 500 g 500g
11400007 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410007 Supplier Thermo Scientific Alfa Aesar Supplier No. B20944.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.

Solubility
Miscible with diethyl ether and acetone. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
MDL Number MFCD00008912
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Melting Point -75°C
Density 0.855
Boiling Point 285°C (25 mmHg)
Flash Point 218°C (424°F)
Assay Percent Range 0.97
Refractive Index 1.4955
Quantity 500 g
Sensitivity Air and light sensitive
Merck Index 14,8768
Solubility Information Miscible with diethyl ether and acetone. Immiscible with water.
Formula Weight 410.73
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Squalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.