missing translation for 'onlineSavingsMsg'
Learn More

Squalene, 99+%, Thermo Scientific Chemicals

Product Code. 11994421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11994421 2.5 L Glass bottle 2.5 L
10226790 100 mL Glass bottle 100mL
10229883 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11994421 Supplier Thermo Scientific Acros Supplier No. 207470025
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Squalene, CAS # 111-02-4, is a biosynthetic precursor to all steroids and ergosterol and is used as an immunologic adjuvant in vaccines.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Squalene is a precursor to steroids and ergosterols
• It can be used in preparation of lipids, steroids, and fat-soluble vitamins

Applications
• Squalene is used as an immunologic adjuvant in vaccines
• It plays a vital role in the preparation of cholesterol, steroid hormones, and vitamin D
• It can also serve as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
MDL Number MFCD00008912
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Acidity 1mg KOH/g max.
Color Colorless to Yellow
Density 0.855g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula {(CH3)2C{=CHCH2CH2C(CH3)}2=CHCH2-}2
Refractive Index 1.4945 to 1.4965 (20°C, 589nm)
Quantity 2.5 L
Saponification Value 1mg KOH/g max.
Formula Weight 410.71
Percent Purity ≥99.0%
Physical Form Oil
Chemical Name or Material Squalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.