missing translation for 'onlineSavingsMsg'
Learn More

Squalene, MP Biomedicals™

Product Code. 11456362 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456362 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456362 Supplier MP Biomedicals Supplier No. 0210294891
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Functions as the biochemical precursor to the whole family of steroids. Squalene, MP Biomedicals is an isoprenoid compound that is structurally similar to beta-carotene and is an intermediate metabolite in the synthesis of cholesterol. Squalene is not ver

  • Functions in the skin as a quencher of singlet oxygen, protecting human skin surface from lipid peroxidation due to exposure to UV and other sources of ionizing radiation
  • Supplementation in mice has resulted in marked increases in cellular and non-specific immune functions in a dose-dependent manner
  • May act as a “sink” for highly lipophilic xenobiotics; since it is a nonpolar substance, it has a higher affinity for un-ionized drugs
  • Supplementation can reduce cholesterol and triglyceride levels; it might be a useful addition to potentiate the effects of some cholesterol-lowering drugs
  • The primary therapeutic use of squalene currently is as an adjunctive therapy in a variety of cancers

Specifications

CAS 111-02-4
Melting Point -75°C (literature)
Density 0.858g/mL at 25°C (Lit.)
Boiling Point 285°C (25mm (literature))
Molecular Formula C30H50
Refractive Index n20/D 1.494 (Lit.)
Quantity 1 L
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
Molecular Weight (g/mol) 410.73
PubChem CID 638072
ChEBI CHEBI:15440
Formula Weight 410.7
Percent Purity ≥98%
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.