missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gosselin™ Square HDPE Bottle, Tamper-Evident Cap
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Pack of 100
Description
- Break-resistant
- Translucent
- Tamper-evident hinged cap
- Assembled
- Sterile
- Graduated
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Material | HDPE |
| No. per Case | 100/Cs |
| Quantity | 100/Cs |
| Sterility | Sterile |
| Graduated | Yes |
| Shape | Square |
| Closure Material | HDPE |
| Closure Type | Tamper-evident Hinged Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction