missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SR 13800
GREENER_CHOICE

Product Code. p-7110824
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18760118

Brand: Tocris Bioscience™ 5431/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent MCT1 inhibitor

SR 13800 is a potent MCT1 inhibitor (IC50 = 0.5 nM); blocks proliferation of Raji lymphoma cells in vitro. Also inhibits lactate uptake in breast cancer cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dihydro-7-[(3-hydroxypropyl)thio]-2-methyl-4-(2-methylpropyl)-6-(1-naphthalenylmethyl)-1H-pyrrolo[3,4-d]pyridazin-1one
CAS 227321-12-2
Quantity 10 mg
Target Monocarboxylate Transporter Inhibitors
Molecular Formula C25H29N3O2S
Formula Weight 435.58
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.