missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SR 7826
GREENER_CHOICE

Product Code. 18784079
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18784079

Brand: Tocris Bioscience™ 5626/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective LIMK inhibitor; antitumor

SR 7826 is a potent and selective LIMK inhibitor (IC50 values are 43, 5536 and 6565 nM for LIMK1, ROCKI and ROCKII, respectively). Inhibits cofilin phosphorylation in A7r5 cells and suppresses migration and invasion of PC-3 cells in vitro. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Hydroxyethyl)-N′-[4,5-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-yl)phenyl]-N-phenylurea
CAS 1219728-20-7
Quantity 50 mg
Target LIMK Inhibitors
Molecular Formula C22H21N5O2
Formula Weight 387.43
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.