missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 9009

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SR 9009 is a Rev-Erbα/β agonist (EC50 = 710 nM; Kd = 800 nM). Alters circadian locomotor activity and core clock hypothalamic gene expression in mice. Induces weight loss in diet-induced obese mice. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-[[[(4-chlorophenyl)methyl][(5-nitro-2-thienyl)methyl]amino]methyl]-1-pyrrolidinecarboxylate |
| CAS | 1379686-30-2 |
| Quantity | 10 mg |
| Target | Nuclear Receptor Agonists |
| Molecular Formula | C20H24ClN3O4S |
| Formula Weight | 437.94 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction