missing translation for 'onlineSavingsMsg'
Learn More

SRT1720 Hydrochloride, TRC

Product Code. 30415725
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415725 50 mg 50mg
30412368 5 mg 5mg
30410190 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415725 Supplier Toronto Research Chemicals Supplier No. TRCS68446050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1001645-58-4
Molecular Formula C25 H23 N7 O S . Cl H
Molecular Weight (g/mol) 506.02
Synonym N-[2-[3-(1-piperazinylmethyl)imidazo[2,1-b]thiazol-6-yl]phenyl]-2-quinoxalinecarboxamide Hydrochloride
IUPAC Name N-[2-[3-(piperazin-1-ylmethyl)imidazo[2,1-b][1,3]thiazol-6-yl]phenyl]quinoxaline-2-carboxamide;hydrochloride
SMILES O=C(NC1=CC=CC=C1C2=CN3C(CN4CCNCC4)=CSC3=N2)C5=NC6=CC=CC=C6N=C5.[xClH]

Specifica

Quantity 50 mg
InChI Formula InChI=1S/C25H23N7OS.ClH/c33-24(22-13-27-20-7-3-4-8-21(20)28-22)29-19-6-2-1-5-18(19)23-15-32-17(16-34-25(32)30-23)14-31-11-9-26-10-12-31;/h1-8,13,15-16,26H,9-12,14H2,(H,29,33);1H
Formula Weight 505.1452
Chemical Name or Material SRT1720 Hydrochloride
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.