Learn More
KD Scientific™ SS Syringe, 1/16 in Swagelok Fitting
Product Code. 18811675
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material Unit Size
18811675 Stainless Steel Each
1 options
Product Code. 18811675 Supplier KD Scientific™ Supplier No. 780806
  • ONLINE EXCLUSIVE PRICE
    2043.80 €
    1684.24 € / Pack of 1
    Save 359.56 €
    17% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Durable, electron-beam construction withstands high force, high pressure applications
  • Eliminates hazards of glass syringe breakage
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge No Needle Included
Latex Free Yes
Disposable Reusable
Volume (Metric) 200 mL
For Use With (Equipment) KD Scientific Syringe Pumps
Connection Type Swagelok
Graduated No
Sterility Non-sterile