missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ ThermoClean™ DC Stabilizing Solution

Product Code. 10449591
Change view
Click to view available options
Color:
Clear
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10449591 Clear 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10449591 Supplier Karl Hecht™ Supplier No. 43170003
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prevents algae/bacterial growth and stops odor formation in water baths. Karl Hecht™ Assistent™ ThermoClean™ DC Stabilizing Solution is suitable for all kinds of thermostats and circulation pumps made of metal, glass or any plastics.

  • Solvent-free
  • Only minor pH-change to the water
  • Prevents foam formation, even with strong circulation
  • No obstructions in tubings and temperature control systems
  • Can also be used for closed temperature control systems
  • Free of aggressive phosphates and oxidizing or reducing substances
  • Available in blue and clear variants 

Specifications

Color Clear
Type Stabilizing Solution
Quantity 100 mL
compliance-icons
Product Identifier
  • Stabilising Solution for Water Baths ThermoClean DC
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: benzyl-C12-16-alkyldimethyl, chlorides,Poly (N,N-Dimethyl-2-hidroxypropyl) ammonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.