Learn More
Hach Lange™ StablCal™ Calibration Set
Product Code. 10763612
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10763612 100 mL Each
1 options
Product Code. 10763612 Supplier Hach Lange™ Supplier No. 2662110
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Type StablCal Calibration Set
Volume (Metric) 100 mL
compliance-icons
Product Identifier
  • Formazin Turbidity Standard 4000 NTU
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P285-In case of inadequate ventilation wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P341-IF INHALED: If breathing is difficult, remove victim to fresh air and keep at rest in a position comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .