missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Stablcal™ Stabilized Formazine Turbidity Standards

Product Code. 10163032
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10163032 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163032 Supplier Hach Lange™ Supplier No. 2660242
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stablcal standards are based on decades of experience in turbidity measurement. With proprietary manufacturing technology, Hach prepares Stablcal Standards in precise concentrations at a wide variety of concentrations - providing accurate turbidimeter calibrations.

  • Stablcal™ Turbidity Standard for all Turbidimeters
  • USEPA accepted for calibration purposes
  • Stablcal™ Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter
  • Stablcal standards are based on decades of experience in turbidity measurement.
  • Precise, Pre-mixed Formazin Solutions
  • Based on Formazin Primary Standards
  • Enhanced Stability Saves Time and Labor
  • Confidence in results
TRUSTED_SUSTAINABILITY

Specifications

Type Turbidity Standard
For Use With (Equipment) Hach Turbidimeters
Turbidity 100 NTU
Volume (Metric) 100 mL
compliance-icons
Product Identifier
  • StablCal Formazin Turbidity Standard; 100 NTU
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: methenamine; hexamethylenetetramine , Sodium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.