missing translation for 'onlineSavingsMsg'
Learn More

Stachydrine, MedChemExpress

Product Code. 16498938
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498938 10 mg 10mg
16489046 100 mg 100mg
16484807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498938 Supplier Medchem Express Supplier No. HYN0298

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Stachydrine is a major constituent of Chinese herb leonurus heterophyllus sweet used to promote blood circulation and dispel blood stasis. Stachydrine can inhibit the NF-κB signal pathway.

Stachydrine is a major constituent of Chinese herb leonurus heterophyllus sweet used to promote blood circulation and dispel blood stasis.

Tekniske data

Chemical Name or Material Stachydrine
CAS 471-87-4
Color White
Purity Grade Notes Research
Molecular Formula C7H13NO2
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (698.42 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (698.42 mM; Need ultrasonic)
SMILES C[N+]1(C)[C@H](C([O-])=O)CCC1
Molecular Weight (g/mol) 143.18
Formula Weight 143.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.