missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Stacking Set for UN30

Product Code. 17520066 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17520066 UN30 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17520066 Supplier Memmert™ Supplier No. UN30B29744
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stacking set for stacking appliances of the same size.

  • For stacking appliances
  • 4 pieces

Specifications

For Use With (Equipment) UN30 Oven
Type Stacking Set
Includes 4 Pieces
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.