missing translation for 'onlineSavingsMsg'
Learn More

BD Stain Droppers

Product Code. 11709032
Change view
Click to view available options
Stain Type:
Dobell and O'Connor's Iodine
Lactophenol Cotton Blue
Methylene Blue
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Stain Type Unit Size
11709032 Lactophenol Cotton Blue Pack of 50
11749032 Dobell and O'Connor's Iodine Pack of 50
11749042 Methylene Blue Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11709032 Supplier BD Supplier No. 261188
 Restricted Item
Estimated Shipment: 15-12-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Identify microorganisms without mixing or measuring stains

To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

  • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
  • Strict quality controls assure uniformity and accuracy

TRUSTED_SUSTAINABILITY

Spécification

Quantity 50 Pack
Certifications/Compliance CE
Format Dropper
For Use With (Application) Used in wet amounts for examination of fungi
Storage Requirements 15° to 30°C
Stain Type Lactophenol Cotton Blue
Volume (Metric) 0.5 mL
Product Type Reagent/Stain Dropper
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • BD BBL Lactophenol Cotton Blue Stain Droppers, 50
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
Conseils de prudence
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informations supplémentaires
  • MIXTURE LIST-Contain: phenol, lactic acid
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.