missing translation for 'onlineSavingsMsg'
Learn More

BD Stain Droppers

Product Code. 11749042
Change view
Click to view available options
Stain Type:
Dobell and O'Connor's Iodine
Lactophenol Cotton Blue
Methylene Blue
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Stain Type unitSize
11749042 Methylene Blue Pack of 50
11749032 Dobell and O'Connor's Iodine Pack of 50
11709032 Lactophenol Cotton Blue Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11749042 Supplier BD Supplier No. 261204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identify microorganisms without mixing or measuring stains

To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

  • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
  • Strict quality controls assure uniformity and accuracy

TRUSTED_SUSTAINABILITY

Specifications

Type Reagent/Stain Dropper
Quantity 50 Pack
Certifications/Compliance CE
Format Dropper
For Use With (Application) Used for presumptive identification of corynebacterium diphtheriae and as an adjunct to the gram stain
Storage Requirements 15° to 30°C
Stain Type Methylene Blue
compliance-icons
Product Identifier
  • BD BBL Methylene Blue Loeffler Stain Droppers
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use CO2, powder, or water spray to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.