missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lamb Pattern Staining Rack

Product Code. 12848735 Shop All Fisherbrand Products
Click to view available options
Holds:
Microscope Slides (25)
Unit Size:
Each

Product Code. 12848735

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Accepts 25 microscope slides measuring 76 x 26mm.

  • Manufactured from stainless steel
  • For use with staining trough MNK-836-K
  • Fitted with swivel handle for easy removal from trough

Specifications

Product Type Staining Rack
Description Lamb pattern
Holds Microscope Slides (25)
Material Stainless Steel
For Use With (Application) Microscope slides sold without Glass rods plated Brass
Quantity 1 Each
Width (Metric) 100 mm
Height (Metric) 25 mm
Depth (Metric) 100 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.