missing translation for 'onlineSavingsMsg'
Learn More

Sampling systems Stainless Spoon 304 SS - 210mm long - 10ml

Brand:  Sampling systems A504-2

Stainless Spoon 304 SS - 210mm long - 10ml

Product Code. 17772229

  • ONLINE EXCLUSIVE PRICE
    45.34 €
    32.39 € / Each
    Save 12.95 €
    28% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.