Learn More
Amsta Rostfritt Stainless Steel Accessory Viton Seal for Chem-Tech Flask 51750-40 and 50 L
Product Code. 30285043
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30285043 None Each
1 options
Product Code. 30285043 Supplier Amsta Rostfritt Supplier No. 50193769
  • ONLINE EXCLUSIVE PRICE
    43.67 €
    37.30 € / Pack of 1
    Save 6.37 €
    14% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest Quality Stainless Steel accessory viton seal for Chem-Tech flask 51750-40 and 50 L. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Container Seal
Capacity (Metric) None
Color Silver
Material Stainless Steel