Learn More
Heinz Herenz Stainless Steel Allergy Lancets with Specially Designed Small Tip
Product Code. 15780158
Change view
Click to view available options
Quantity:
200 Lancets
Unit Size:
Pack of 200
Product Code. Quantity Unit Size
15780158 200 Lancets Pack of 200
1 options
Product Code. 15780158 Supplier Heinz Herenz Supplier No. 1110106
  • ONLINE EXCLUSIVE PRICE
    13.49 €
    9.89 € / Pack of 200
    Save 3.60 €
    26% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Allergy lancets made in Germany. Stainless steel, with specially designed small tip.
Our blood and allergy lancets are made of stainless chrome steel and are nickelfree.
  • Each lancet is individually sterile packed so they are quick and easy to use by hand.
  • The blood lancet ensures excellent blood flow.
  • It also has an approximately 2.9 mm long, asymmetrical tip, which simplifies blood collection and reduces pain significantly.
  • The tip of the allergy lancet is particularly fine, approximately 0.9 mm. This prevents unwanted bleeding.
  • All lancets have a transparent sealing foil. This offers optimal safety when unpacking, as the tip is visible.
Please note:
  • Only suitable for single use
  • Store at a temperature of 15°C to 30°C, with a relative humidity of 40 to 70%
  • The blood lancets are gamma sterilized and sterile according to DIN EN 5561.
Specifications
Material Stainless Chrome Steel
Quantity 200 Lancets
Sterility Sterile
Product Type Lancet