Learn More
Masterflex™ Stainless Steel Barbed Fitting NPT Male Pipe Adapter
Product Code. 11811073
Click to view available options
Thread Size:
1.5 in. NPT(M)
1/2 in. NPT(M)
1/4 in. NPT(M)
3/8 in. NPT(M)
Tubing Size:
1 1/2 in. I.D.
1/16 in. I.D.
1/2 in. I.D.
1/4 in. I.D
1/4 in. I.D.
3/8 in. I.D.
Unit Size:
Each
Product Code. 11811073
Brand: Masterflex™ 3121030
Only null left
Add to basket
Add to basket
Designed for quick and easy assembly without tools
  • Available in a variety of materials and configurations to meet your need
  • Adapts from a NPT male thread to a hose barb
Specifications
Material Stainless Steel
Overall Length (English) 1 27/32 in.
Tubing Size 1/4 in. I.D
Hose Barb 1/4 in.
Outlet Port 1/2 in. NPT(M)
Port Size 1/4 in.
Thread Size 1/2 in. NPT(M)
Width (English) 7/8 in.
Product Type Barbed Fitting NPT Male Pipe Adapter