Learn More
Scientific Industries Stainless Steel Beads
Product Code. 17201762 Shop All Ohaus Products
Change view
Click to view available options
Diameter (Metric) Probe:
2.4 mm
3.2 mm
5.0 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Probe Unit Size
17201762 3.2 mm Pack of 100
17291752 2.4 mm Pack of 100
17211762 5.0 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17201762 Supplier Scientific Industries Supplier No. 83041299
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless Steel Beads come in a variety of sizes and are designed for use in bead beating procedures with Bead Genie bead beater
  • Can be cleaned and reused
TRUSTED_SUSTAINABILITY
Specifications
Product Type Beads
Diameter (English) Probe 0.1259 in.
Diameter (Metric) Probe 3.2 mm
Material Stainless Steel
For Use With (Application) Bead Beating
For Use With (Equipment) Bead Genie
Model SI-BS03
Packaging Quantity 100 Pk.